Product Details

SNP ID
rs72876670
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:32020434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGATCCTGTCTCAAAAAAAAAA[C/T]GTGCTTACCGTATCGTTCAACAGTT
Phenotype
MIM: 602489
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KHDRBS1 PubMed Links

Gene Details

Gene
KHDRBS1
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271878.1 Intron NP_001258807.1
NM_006559.2 Intron NP_006550.1

View Full Product Details