Product Details

SNP ID
rs72682004
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:75203951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCCTTTTGTTGTTTTTTTTTT[G/T]TTGTTGTTTTTTAAACAGCATAGTT
Phenotype
MIM: 604425
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LHX8 PubMed Links

Gene Details

Gene
LHX8
Gene Name
LIM homeobox 8
There are no transcripts associated with this gene.

Gene
SLC44A5
Gene Name
solute carrier family 44 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130058.1 4938 Intron NP_001123530.1
NM_001320283.1 4938 Intron NP_001307212.1
NM_001320285.1 4938 Intron NP_001307214.1
NM_001320287.1 4938 Intron NP_001307216.1
NM_152697.4 4938 Intron NP_689910.2
XM_006710445.3 4938 UTR 3 XP_006710508.1
XM_011540986.2 4938 UTR 3 XP_011539288.1
XM_011540987.2 4938 UTR 3 XP_011539289.1
XM_017000607.1 4938 UTR 3 XP_016856096.1
XM_017000608.1 4938 Intron XP_016856097.1
XM_017000609.1 4938 UTR 3 XP_016856098.1
XM_017000610.1 4938 UTR 3 XP_016856099.1
XM_017000611.1 4938 UTR 3 XP_016856100.1

View Full Product Details