Product Details

SNP ID
rs74108028
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:102877315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATGCATAATATTTTTGGCTGA[A/G]ATTCAGCAAAATGACTGTCGGCAGA
Phenotype
MIM: 120280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL11A1 PubMed Links

Gene Details

Gene
COL11A1
Gene Name
collagen type XI alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190709.1 6443 UTR 3 NP_001177638.1
NM_001854.3 6443 UTR 3 NP_001845.3
NM_080629.2 6443 UTR 3 NP_542196.2
NM_080630.3 6443 UTR 3 NP_542197.3
XM_017000334.1 6443 UTR 3 XP_016855823.1
XM_017000335.1 6443 Intron XP_016855824.1
XM_017000336.1 6443 Intron XP_016855825.1
XM_017000337.1 6443 Intron XP_016855826.1

View Full Product Details