Product Details

SNP ID
rs73615728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125096515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTTCTACTTTAAAGTGGGGA[A/T]GATAAAAATGAGACCGTTGTAAAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC37A2 PubMed Links

Gene Details

Gene
SLC37A2
Gene Name
solute carrier family 37 member 2
There are no transcripts associated with this gene.

Gene
TMEM218
Gene Name
transmembrane protein 218
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080546.2 2071 UTR 3 NP_001074015.1
NM_001258238.1 2071 UTR 3 NP_001245167.1
NM_001258239.1 2071 UTR 3 NP_001245168.1
NM_001258240.1 2071 UTR 3 NP_001245169.1
NM_001258241.1 2071 UTR 3 NP_001245170.1
NM_001258242.1 2071 UTR 3 NP_001245171.1
NM_001258243.1 2071 UTR 3 NP_001245172.1
NM_001258244.1 2071 UTR 3 NP_001245173.1
NM_001258245.1 2071 UTR 3 NP_001245174.1
NM_001258246.1 2071 UTR 3 NP_001245175.1
NM_001258247.1 2071 UTR 3 NP_001245176.1
XM_006718779.3 2071 UTR 3 XP_006718842.1
XM_006718780.3 2071 UTR 3 XP_006718843.1
XM_006718781.3 2071 UTR 3 XP_006718844.1
XM_006718783.3 2071 UTR 3 XP_006718846.1
XM_017017322.1 2071 UTR 3 XP_016872811.1
XM_017017323.1 2071 UTR 3 XP_016872812.1
XM_017017324.1 2071 UTR 3 XP_016872813.1

View Full Product Details