Product Details

SNP ID
rs72916996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60915470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCTATGCATCCGAAATTCCTG[A/C]GGAGCTTTGCAAAACAAAGGGCTGA
Phenotype
MIM: 608330
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRPF19 PubMed Links

Gene Details

Gene
PRPF19
Gene Name
pre-mRNA processing factor 19
There are no transcripts associated with this gene.

Gene
TMEM109
Gene Name
transmembrane protein 109
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024092.2 Intron NP_076997.1
Gene
TMEM132A
Gene Name
transmembrane protein 132A
There are no transcripts associated with this gene.

View Full Product Details