Product Details

SNP ID
rs73502133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62665312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAGCTCCGCGGTGCGGGAGGC[C/T]TTTCGGAGGGTGGTGAGCTAGTAAG
Phenotype
MIM: 616097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf98 PubMed Links

Gene Details

Gene
C11orf98
Gene Name
chromosome 11 open reading frame 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286086.1 61 Intron NP_001273015.1
Gene
LBHD1
Gene Name
LBH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024099.3 61 Intron NP_077004.2
Gene
METTL12
Gene Name
methyltransferase like 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043229.1 61 UTR 5 NP_001036694.1
XM_005274232.4 61 UTR 5 XP_005274289.1
Gene
SNORA57
Gene Name
small nucleolar RNA, H/ACA box 57
There are no transcripts associated with this gene.

Gene
UQCC3
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 3
There are no transcripts associated with this gene.

View Full Product Details