Product Details

SNP ID
rs72924108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64302069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGTGTACGTCTTTCCATTAG[C/T]CAGTTGCTGGTTACAATTTCTAAGC
Phenotype
MIM: 601998 MIM: 605720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRRA PubMed Links

Gene Details

Gene
ESRRA
Gene Name
estrogen related receptor alpha
There are no transcripts associated with this gene.

Gene
KCNK4
Gene Name
potassium two pore domain channel subfamily K member 4
There are no transcripts associated with this gene.

Gene
KCNK4-TEX40
Gene Name
KCNK4-TEX40 readthrough
There are no transcripts associated with this gene.

Gene
TEX40
Gene Name
testis expressed 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039496.1 Intron NP_001034585.1

View Full Product Details