Product Details

SNP ID
rs67719472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:76687491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTGTCCTGTCCTGAGCCCCT[C/T]CTGCAGAGATTCTGTGTGTGTGTGT
Phenotype
MIM: 601138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCY2EP PubMed Links

Gene Details

Gene
GUCY2EP
Gene Name
guanylate cyclase 2E, pseudogene
There are no transcripts associated with this gene.

View Full Product Details