Product Details

SNP ID
rs73534053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70862815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGATCCTTTGGGATTCCCCCA[A/C]GGGAGTAGTCAACTGGGCAGCAGGA
Phenotype
MIM: 603290
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR3664 PubMed Links

Gene Details

Gene
MIR3664
Gene Name
microRNA 3664
There are no transcripts associated with this gene.

Gene
SHANK2
Gene Name
SH3 and multiple ankyrin repeat domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012309.4 Intron NP_036441.2
NM_133266.4 Intron NP_573573.2
XM_005277932.3 Intron XP_005277989.1
XM_017017387.1 Intron XP_016872876.1
XM_017017388.1 Intron XP_016872877.1
XM_017017389.1 Intron XP_016872878.1
XM_017017390.1 Intron XP_016872879.1
Gene
SHANK2-AS3
Gene Name
SHANK2 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details