Product Details

SNP ID
rs73241728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:113432838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTAAATTAGCTACAAAATCAA[C/T]TACAAGGAGAATGATTAAATATAAA
Phenotype
MIM: 159350 MIM: 610709
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCC PubMed Links

Gene Details

Gene
MCC
Gene Name
mutated in colorectal cancers
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085377.1 2194 Intron NP_001078846.1
NM_002387.2 2194 Intron NP_002378.1
XM_005271991.2 2194 Intron XP_005272048.1
XM_017009473.1 2194 Intron XP_016864962.1
XM_017009474.1 2194 Intron XP_016864963.1
Gene
TSSK1B
Gene Name
testis specific serine kinase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032028.3 2194 UTR 3 NP_114417.1

View Full Product Details