Product Details

SNP ID
rs73260551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115615478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATTTTCCTATTTCATTTGCC[C/G]CCATTGTAGTGTCTTAAATTATTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927100 PubMed Links

Gene Details

Gene
LOC101927100
Gene Name
uncharacterized LOC101927100
There are no transcripts associated with this gene.

Gene
TMED7
Gene Name
transmembrane p24 trafficking protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181836.5 1793 UTR 3 NP_861974.1
Gene
TMED7-TICAM2
Gene Name
TMED7-TICAM2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164468.3 1793 Intron NP_001157940.1
NM_001164469.3 1793 Intron NP_001157941.1

View Full Product Details