Product Details

SNP ID
rs73793010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:119631621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACAGAGAAAGAGTCACACACCT[C/T]CACACACAGTGGATATAGTTATATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM170A PubMed Links

Gene Details

Gene
FAM170A
Gene Name
family with sequence similarity 170 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163991.1 Intron NP_001157463.1
NM_182761.3 Intron NP_877438.2

View Full Product Details