Product Details

SNP ID
rs73262392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131280495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGAAAGTGAAAATTCTGTTTGA[C/T]GAGGATAAAGCTGTAGGTAAAGTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42SE2 PubMed Links

Gene Details

Gene
CDC42SE2
Gene Name
CDC42 small effector 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038702.1 Intron NP_001033791.1
NM_020240.2 Intron NP_064625.1
XM_017009648.1 Intron XP_016865137.1
XM_017009649.1 Intron XP_016865138.1

View Full Product Details