Product Details

SNP ID
rs73263483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138872820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGCAAATTAATGTGAGCATTG[A/T]TTCATTTAACACTTAAAATATGGTT
Phenotype
MIM: 116805 MIM: 610868
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTNNA1 PubMed Links

Gene Details

Gene
CTNNA1
Gene Name
catenin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290307.2 2198 Intron NP_001277236.1
NM_001290309.2 2198 Intron NP_001277238.1
NM_001290310.2 2198 Intron NP_001277239.1
NM_001290312.1 2198 Intron NP_001277241.1
NM_001323982.1 2198 Intron NP_001310911.1
NM_001323983.1 2198 Intron NP_001310912.1
NM_001323984.1 2198 Intron NP_001310913.1
NM_001323985.1 2198 Intron NP_001310914.1
NM_001323986.1 2198 Intron NP_001310915.1
NM_001323987.1 2198 Intron NP_001310916.1
NM_001323988.1 2198 Intron NP_001310917.1
NM_001323989.1 2198 Intron NP_001310918.1
NM_001323990.1 2198 Intron NP_001310919.1
NM_001323991.1 2198 Intron NP_001310920.1
NM_001323992.1 2198 Intron NP_001310921.1
NM_001323993.1 2198 Intron NP_001310922.1
NM_001323994.1 2198 Intron NP_001310923.1
NM_001323995.1 2198 Intron NP_001310924.1
NM_001323996.1 2198 Intron NP_001310925.1
NM_001323997.1 2198 Intron NP_001310926.1
NM_001323998.1 2198 Intron NP_001310927.1
NM_001323999.1 2198 Intron NP_001310928.1
NM_001324000.1 2198 Intron NP_001310929.1
NM_001324001.1 2198 Intron NP_001310930.1
NM_001324002.1 2198 Intron NP_001310931.1
NM_001324003.1 2198 Intron NP_001310932.1
NM_001324004.1 2198 Intron NP_001310933.1
NM_001324005.1 2198 Intron NP_001310934.1
NM_001324006.1 2198 Intron NP_001310935.1
NM_001324007.1 2198 Intron NP_001310936.1
NM_001324008.1 2198 Intron NP_001310937.1
NM_001324009.1 2198 Intron NP_001310938.1
NM_001324010.1 2198 Intron NP_001310939.1
NM_001324011.1 2198 Intron NP_001310940.1
NM_001324012.1 2198 Intron NP_001310941.1
NM_001324013.1 2198 Intron NP_001310942.1
NM_001903.4 2198 Intron NP_001894.2
Gene
LRRTM2
Gene Name
leucine rich repeat transmembrane neuronal 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015564.2 2198 UTR 3 NP_056379.1

View Full Product Details