Product Details

SNP ID
rs73197879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:103086804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCTTTATAAGAATGAGGTACAC[A/G]TTTGACTCATCTTTTACTTTTCCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIN1 PubMed Links

Gene Details

Gene
GIN1
Gene Name
gypsy retrotransposon integrase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317954.1 2551 UTR 3 NP_001304883.1
NM_017676.2 2551 UTR 3 NP_060146.2
XM_005272028.3 2551 UTR 3 XP_005272085.1
XM_017009599.1 2551 Intron XP_016865088.1

View Full Product Details