Product Details

SNP ID
rs73348572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2666887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTTGCCCCCACCTCTGGTGTC[C/T]GATTGAGATTTCTTCACCACCATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYLK4 PubMed Links

Gene Details

Gene
MYLK4
Gene Name
myosin light chain kinase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249078.3 2399 UTR 3 XP_005249135.2
XM_005249079.2 2399 UTR 3 XP_005249136.1
XM_006715082.3 2399 UTR 3 XP_006715145.1
XM_011514567.2 2399 UTR 3 XP_011512869.1
XM_017010819.1 2399 UTR 3 XP_016866308.1

View Full Product Details