Product Details

SNP ID
rs73552805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132893502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGCAGGGACTCCGGAGCTCAT[C/T]TGCTCTGCGTCTTCCTATCCCCCTC
Phenotype
MIM: 605284
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPACA9 PubMed Links

Gene Details

Gene
SPACA9
Gene Name
sperm acrosome associated 9
There are no transcripts associated with this gene.

Gene
TSC1
Gene Name
tuberous sclerosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000368.4 6507 UTR 3 NP_000359.1
NM_001162426.1 6507 UTR 3 NP_001155898.1
NM_001162427.1 6507 UTR 3 NP_001155899.1
XM_005272211.1 6507 UTR 3 XP_005272268.1
XM_006717271.1 6507 UTR 3 XP_006717334.1
XM_011518979.2 6507 UTR 3 XP_011517281.1
XM_017015096.1 6507 UTR 3 XP_016870585.1
XM_017015097.1 6507 UTR 3 XP_016870586.1
XM_017015098.1 6507 UTR 3 XP_016870587.1
XM_017015099.1 6507 UTR 3 XP_016870588.1
XM_017015100.1 6507 UTR 3 XP_016870589.1
XM_017015101.1 6507 UTR 3 XP_016870590.1

View Full Product Details