Product Details

SNP ID
rs74045379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:452282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACCGTTCTACGGGACATAGCAGA[G/T]AACTAAACAAGAAGCTTAGATTCTG
Phenotype
MIM: 612793
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTDSS2 PubMed Links

Gene Details

Gene
PTDSS2
Gene Name
phosphatidylserine synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005253153.1 775 Intron XP_005253210.1
XM_011520391.2 775 UTR 5 XP_011518693.1
XM_011520392.2 775 Intron XP_011518694.1
XM_011520393.2 775 Intron XP_011518695.1
XM_017018368.1 775 UTR 5 XP_016873857.1
XM_017018369.1 775 Intron XP_016873858.1
XM_017018370.1 775 Intron XP_016873859.1
XM_017018371.1 775 Intron XP_016873860.1

View Full Product Details