Product Details

SNP ID
rs73470740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100402212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGACTGCCACCGCCCTGTGGA[A/G]ATGAGGCACCTAAGCTTCAGGATGG
Phenotype
MIM: 615276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERS3 PubMed Links

Gene Details

Gene
CERS3
Gene Name
ceramide synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290341.2 2086 UTR 3 NP_001277270.1
NM_001290342.2 2086 UTR 3 NP_001277271.1
NM_001290343.1 2086 UTR 3 NP_001277272.1
NM_178842.4 2086 UTR 3 NP_849164.2
XM_011521355.2 2086 UTR 3 XP_011519657.1
XM_011521357.2 2086 UTR 3 XP_011519659.1
XM_011521358.1 2086 UTR 3 XP_011519660.1
XM_017022002.1 2086 UTR 3 XP_016877491.1
XM_017022003.1 2086 UTR 3 XP_016877492.1
XM_017022004.1 2086 UTR 3 XP_016877493.1
Gene
CERS3-AS1
Gene Name
CERS3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details