Product Details

SNP ID
rs67616875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:100603326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTCTGTAAAGGTAATGAGCCAG[A/C]CCTCCCCTCCCCCGTTGTTTATTCC
Phenotype
MIM: 615052 MIM: 610350
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASB7 PubMed Links

Gene Details

Gene
ASB7
Gene Name
ankyrin repeat and SOCS box containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024708.3 Intron NP_078984.2
NM_198243.2 Intron NP_937886.1
Gene
LINS1
Gene Name
lines homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040616.2 Intron NP_001035706.1
XM_005254941.1 Intron XP_005254998.1
XM_005254943.1 Intron XP_005255000.1
XM_017022398.1 Intron XP_016877887.1
XM_017022399.1 Intron XP_016877888.1
XM_017022400.1 Intron XP_016877889.1

View Full Product Details