Product Details
- SNP ID
-
rs72613115
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1406384 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCGGTGCGGGCCCCGAACCTGGC[C/T]GGGGAGGCGGCGGTAGCGCGGGAAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NKX1-1
PubMed Links
Gene Details
- Gene
- NKX1-1
- Gene Name
- NK1 homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290079.1 |
59 |
Missense Mutation |
CAG,CGG |
Q20R |
NP_001277008.1 |
View Full Product Details