Product Details

SNP ID
rs72496213
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:12132532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATGGACTCCTGATCTGTCGGT[G/T]TCGTCGAAGAGAGGTTTAGCAGGGA
Phenotype
MIM: 610186
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
FAM66D PubMed Links
Additional Information
For this assay, SNP(s) [rs373078712] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM66D
Gene Name
family with sequence similarity 66 member D
There are no transcripts associated with this gene.

Gene
LOC392196
Gene Name
ubiquitin specific peptidase 17-like family member 2 pseudogene
There are no transcripts associated with this gene.

Gene
USP17L2
Gene Name
ubiquitin specific peptidase 17-like family member 2
There are no transcripts associated with this gene.

Gene
USP17L7
Gene Name
ubiquitin specific peptidase 17-like family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256869.1 Intron NP_001243798.1

View Full Product Details