Product Details

SNP ID
rs72760937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1063264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAGGACTCAGACAAAACAAGGC[A/C]CAAGAAAAGCCTGCTCTCTGTGTCT
Phenotype
MIM: 604055
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IDI1 PubMed Links

Gene Details

Gene
IDI1
Gene Name
isopentenyl-diphosphate delta isomerase 1
There are no transcripts associated with this gene.

Gene
WDR37
Gene Name
WD repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014023.3 Intron NP_054742.2

View Full Product Details