Product Details
- SNP ID
-
rs73615943
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5369062 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCGCAGGCTGTGTTTGAACCGC[A/G]CCTTCACATGAGTCCAAAGCACCTG
- Phenotype
-
MIM: 605901
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UCN3
PubMed Links
Gene Details
- Gene
- UCN3
- Gene Name
- urocortin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_053049.3 |
|
Intron |
|
|
NP_444277.2 |
View Full Product Details