Product Details

SNP ID
rs73599177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16591370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATTGCAGTTTGTGGGGATCTC[A/G]GATAATTTCCTCAGGATACATTCCT
Phenotype
MIM: 179555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSU1 PubMed Links

Gene Details

Gene
RSU1
Gene Name
Ras suppressor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012425.3 3001 UTR 3 NP_036557.1
NM_152724.2 3001 UTR 3 NP_689937.2
XM_005252552.3 3001 UTR 3 XP_005252609.1

View Full Product Details