Product Details

SNP ID
rs74122558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13167902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGAGCCACTGGGGATCCGAGAAA[C/T]GTGCTGATAGTAGATTTCACATCCC
Phenotype
MIM: 609357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM10 PubMed Links

Gene Details

Gene
MCM10
Gene Name
minichromosome maintenance 10 replication initiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018518.4 Intron NP_060988.3
NM_182751.2 Intron NP_877428.1
XM_011519538.2 Intron XP_011517840.1

View Full Product Details