Product Details

SNP ID
rs74121340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13646042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCATGGACCGGGGCTAACAG[A/T]ACCCTCTACGACTCCCACAGGTCTC
Phenotype
MIM: 616305 MIM: 604993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FRMD4A PubMed Links

Gene Details

Gene
FRMD4A
Gene Name
FERM domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318336.1 4517 UTR 3 NP_001305265.1
NM_001318337.1 4517 UTR 3 NP_001305266.1
NM_001318338.1 4517 UTR 3 NP_001305267.1
NM_018027.4 4517 UTR 3 NP_060497.3
XM_005252489.2 4517 UTR 3 XP_005252546.1
XM_005252490.2 4517 UTR 3 XP_005252547.1
XM_005252495.4 4517 UTR 3 XP_005252552.1
XM_006717457.2 4517 UTR 3 XP_006717520.1
XM_006717458.3 4517 UTR 3 XP_006717521.1
XM_006717459.2 4517 UTR 3 XP_006717522.1
XM_006717460.2 4517 UTR 3 XP_006717523.1
XM_006717464.1 4517 UTR 3 XP_006717527.1
XM_011519539.1 4517 UTR 3 XP_011517841.1
XM_011519540.1 4517 UTR 3 XP_011517842.1
XM_011519541.1 4517 UTR 3 XP_011517843.1
XM_011519543.2 4517 UTR 3 XP_011517845.1
XM_017016393.1 4517 UTR 3 XP_016871882.1
XM_017016394.1 4517 UTR 3 XP_016871883.1
XM_017016395.1 4517 UTR 3 XP_016871884.1
XM_017016396.1 4517 UTR 3 XP_016871885.1
Gene
LOC101928524
Gene Name
uncharacterized LOC101928524
There are no transcripts associated with this gene.

Gene
PRPF18
Gene Name
pre-mRNA processing factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003675.3 4517 Intron NP_003666.1
XM_011519754.2 4517 Intron XP_011518056.1
XM_017016860.1 4517 Intron XP_016872349.1

View Full Product Details