Product Details

SNP ID
rs66913836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113760418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCCTTAATGTTTAAAAATAGC[A/C]TCTGACTTAGATTTTCTTCTGACAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00552 PubMed Links

Gene Details

Gene
LINC00552
Gene Name
long intergenic non-protein coding RNA 552
There are no transcripts associated with this gene.

Gene
LOC105377805
Gene Name
basic salivary proline-rich protein 4-like
There are no transcripts associated with this gene.

Gene
TMEM255B
Gene Name
transmembrane protein 255B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182614.2 Intron NP_872420.1
XM_005272413.3 Intron XP_005272470.1
XM_005272415.2 Intron XP_005272472.1
XM_006719986.2 Intron XP_006720049.1
XM_017020558.1 Intron XP_016876047.1
XM_017020559.1 Intron XP_016876048.1
XM_017020560.1 Intron XP_016876049.1

View Full Product Details