Product Details

SNP ID
rs73291809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37505554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCATCTGAAGCACTGTTGTGG[C/T]CATTTTCTGCTGTTGACTGAACATG
Phenotype
MIM: 606618 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007026.3 Intron NP_008957.1
XM_005256977.3 Intron XP_005257034.1
XM_011524234.1 Intron XP_011522536.1
Gene
SYNRG
Gene Name
synergin, gamma
There are no transcripts associated with this gene.

View Full Product Details