Product Details

SNP ID
rs62190274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241655641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTGGGCAGCTGAAGCTAGTCT[A/G]CAGCCATCTGTGCGGGTGCCGGGCC
Phenotype
MIM: 611338
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG4B PubMed Links

Gene Details

Gene
ATG4B
Gene Name
autophagy related 4B cysteine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013325.4 Intron NP_037457.3
NM_178326.2 Intron NP_847896.1
XM_005246992.3 Intron XP_005247049.2
XM_005246993.2 Intron XP_005247050.2
XM_005246995.3 Intron XP_005247052.1
XM_005246996.2 Intron XP_005247053.1
XM_017003637.1 Intron XP_016859126.1
XM_017003638.1 Intron XP_016859127.1
XM_017003639.1 Intron XP_016859128.1
XM_017003640.1 Intron XP_016859129.1

View Full Product Details