Product Details

SNP ID
rs79585663
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:927758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGGAGGTGCTGTCAGGGGCCG[A/C]GTTTTCAGATCTGTGTGCTGTGGCC
Phenotype
MIM: 616765
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130417 PubMed Links

Gene Details

Gene
LOC100130417
Gene Name
uncharacterized LOC100130417
There are no transcripts associated with this gene.

Gene
LOC107985728
Gene Name
collagen alpha-1(III) chain-like
There are no transcripts associated with this gene.

Gene
LOC284600
Gene Name
uncharacterized LOC284600
There are no transcripts associated with this gene.

Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 Intron NP_689699.2

View Full Product Details