Product Details
- SNP ID
-
rs76790197
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:160092608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCGCAGTCCTGGGGGGCCACCC[C/T]GCACAGAGATGTTGCACAGCAGGGA
- Phenotype
-
MIM: 606644
MIM: 600932
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IGSF8
PubMed Links
Gene Details
- Gene
- IGSF8
- Gene Name
- immunoglobulin superfamily member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206665.2 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
NP_001193594.1 |
NM_001320247.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
NP_001307176.1 |
NM_052868.5 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
NP_443100.1 |
XM_006711631.3 |
1481 |
Missense Mutation |
CAG,CGG |
Q450R |
XP_006711694.1 |
XM_011510159.2 |
1481 |
Missense Mutation |
CAG,CGG |
Q445R |
XP_011508461.1 |
XM_017002835.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
XP_016858324.1 |
XM_017002836.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
XP_016858325.1 |
XM_017002837.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q467R |
XP_016858326.1 |
XM_017002838.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q445R |
XP_016858327.1 |
XM_017002839.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q445R |
XP_016858328.1 |
XM_017002840.1 |
1481 |
Missense Mutation |
CAG,CGG |
Q445R |
XP_016858329.1 |
- Gene
- KCNJ9
- Gene Name
- potassium voltage-gated channel subfamily J member 9
There are no transcripts associated with this gene.
View Full Product Details