Product Details

SNP ID
rs78390484
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:160289206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATTATTACTGGAAGGAAGATAT[A/G]TACTTTGGTGGGGTGCCTGCATGGG
Phenotype
MIM: 601924 MIM: 600279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 5047 UTR 3 NP_001091868.1
NM_004371.3 5047 UTR 3 NP_004362.2
Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
There are no transcripts associated with this gene.

View Full Product Details