Product Details

SNP ID
rs77068976
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:166848955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCCAGAAAATGAAGAATCTGAC[A/G]TAAAGCCTCCAGACTGGCCAAACCC
Phenotype
MIM: 612763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGK PubMed Links

Gene Details

Gene
POGK
Gene Name
pogo transposable element with KRAB domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314014.1 856 Missense Mutation ATA,GTA I8V NP_001300943.1
NM_017542.4 856 Missense Mutation ATA,GTA I126V NP_060012.3
XM_005245370.3 856 Missense Mutation ATA,GTA I126V XP_005245427.1
XM_011509814.2 856 Missense Mutation ATA,GTA I126V XP_011508116.1
XM_011509815.1 856 Missense Mutation ATA,GTA I126V XP_011508117.1
XM_017001914.1 856 Missense Mutation ATA,GTA I126V XP_016857403.1
XM_017001915.1 856 Missense Mutation ATA,GTA I8V XP_016857404.1
Gene
TADA1
Gene Name
transcriptional adaptor 1
There are no transcripts associated with this gene.

View Full Product Details