Product Details
- SNP ID
-
rs76170587
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:167054049 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCCAGGAGCTCCTGCCAACTAC[A/G]AGGGAAGTGGAGGCTGCAGCCTGGT
- Phenotype
-
MIM: 602171
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPA33
PubMed Links
Gene Details
- Gene
- GPA33
- Gene Name
- glycoprotein A33
View Full Product Details