Product Details

SNP ID
rs80276389
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:169465974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCAAAATAACTGGCATAGATCA[A/G]AAACTAGAAGGGGGAAAAGCAGTTT
Phenotype
MIM: 603941
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC181 PubMed Links

Gene Details

Gene
CCDC181
Gene Name
coiled-coil domain containing 181
There are no transcripts associated with this gene.

Gene
SLC19A2
Gene Name
solute carrier family 19 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319667.1 970 Silent Mutation CTG,TTG L256L NP_001306596.1
NM_006996.2 970 Silent Mutation CTG,TTG L457L NP_008927.1

View Full Product Details