Product Details
- SNP ID
-
rs76510731
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:169513937 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTGTGAAAGTTATCCAGGTCTA[A/T]CAATTATTATTTAAAGTAATATCTG
- Phenotype
-
MIM: 612309
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
F5
PubMed Links
Gene Details
- Gene
- F5
- Gene Name
- coagulation factor V
View Full Product Details