Product Details

SNP ID
rs78940115
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:173799681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTTTGAGGGGATTTTGATTGCC[A/G]GTTCTCCGCAGTGAAATGCTAGGAA
Phenotype
MIM: 611503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPL PubMed Links

Gene Details

Gene
CENPL
Gene Name
centromere protein L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127181.2 2405 UTR 3 NP_001120653.1
NM_001171182.1 2405 UTR 3 NP_001164653.1
NM_033319.3 2405 UTR 3 NP_201576.1

View Full Product Details