Product Details

SNP ID
rs78631664
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:151292483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCACTTCCTGGGCCCCAGAA[C/T]GGAATAAGAGGATGGCCTGTGGAAA
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI4KB PubMed Links

Gene Details

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198774.1 3248 UTR 3 NP_001185703.1
XM_005245264.3 3248 UTR 3 XP_005245321.1
XM_011509634.1 3248 UTR 3 XP_011507936.1
XM_011509635.1 3248 UTR 3 XP_011507937.1
XM_011509636.2 3248 UTR 3 XP_011507938.1
XM_011509637.2 3248 UTR 3 XP_011507939.1
XM_011509638.1 3248 UTR 3 XP_011507940.1
XM_017001486.1 3248 UTR 3 XP_016856975.1
XM_017001487.1 3248 UTR 3 XP_016856976.1
XM_017001488.1 3248 UTR 3 XP_016856977.1
XM_017001489.1 3248 UTR 3 XP_016856978.1
XM_017001490.1 3248 UTR 3 XP_016856979.1
XM_017001491.1 3248 UTR 3 XP_016856980.1
XM_017001492.1 3248 UTR 3 XP_016856981.1
Gene
ZNF687
Gene Name
zinc finger protein 687
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304763.1 3248 Intron NP_001291692.1
NM_001304764.1 3248 Intron NP_001291693.1
NM_020832.2 3248 Intron NP_065883.1
XM_005245366.3 3248 Intron XP_005245423.1
XM_011509811.2 3248 Intron XP_011508113.1
XM_011509812.2 3248 Intron XP_011508114.1
XM_011509813.2 3248 Intron XP_011508115.1

View Full Product Details