Product Details

SNP ID
rs79029362
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:178547273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCATAAGTCACTTACTGGGTT[G/T]GTCCTCCTTGAGTGGACTGTGTTGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf220 PubMed Links

Gene Details

Gene
C1orf220
Gene Name
chromosome 1 open reading frame 220
There are no transcripts associated with this gene.

View Full Product Details