Product Details

SNP ID
rs74507040
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:179740021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCAGTCTCTTTAACAACTAAG[C/T]ATGTGACTACCATACGATCTAGCAA
Phenotype
MIM: 611727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM163A PubMed Links

Gene Details

Gene
FAM163A
Gene Name
family with sequence similarity 163 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173509.2 Intron NP_775780.1
XM_005244913.2 Intron XP_005244970.1
XM_006711174.2 Intron XP_006711237.1
XM_006711175.2 Intron XP_006711238.1
XM_006711176.2 Intron XP_006711239.1
XM_006711177.1 Intron XP_006711240.1
XM_011509217.1 Intron XP_011507519.1
XM_011509221.1 Intron XP_011507523.1
XM_011509226.1 Intron XP_011507528.1
XM_011509228.1 Intron XP_011507530.1
XM_017000386.1 Intron XP_016855875.1
XM_017000387.1 Intron XP_016855876.1
XM_017000388.1 Intron XP_016855877.1
XM_017000389.1 Intron XP_016855878.1
XM_017000390.1 Intron XP_016855879.1
XM_017000391.1 Intron XP_016855880.1
XM_017000392.1 Intron XP_016855881.1
XM_017000393.1 Intron XP_016855882.1

View Full Product Details