Product Details

SNP ID
rs78432452
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:151770285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGATCTGCTGTTCTTCCTTGCTT[G/T]TCGCGCTGAGGCAGCTGGATAACTC
Phenotype
MIM: 611824 MIM: 605138 MIM: 609501
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MRPL9 PubMed Links

Gene Details

Gene
MRPL9
Gene Name
mitochondrial ribosomal protein L9
There are no transcripts associated with this gene.

Gene
OAZ3
Gene Name
ornithine decarboxylase antizyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134939.1 2227 Intron NP_001128411.1
NM_001301371.1 2227 Intron NP_001288300.1
NM_016178.2 2227 Intron NP_057262.2
Gene
TDRKH
Gene Name
tudor and KH domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083963.1 2227 Intron NP_001077432.1
NM_001083964.1 2227 Intron NP_001077433.1
NM_001083965.1 2227 Intron NP_001077434.1
NM_006862.3 2227 Intron NP_006853.2
XM_005244856.4 2227 Intron XP_005244913.1
XM_017000122.1 2227 Intron XP_016855611.1
XM_017000123.1 2227 UTR 3 XP_016855612.1
XM_017000124.1 2227 Intron XP_016855613.1
XM_017000125.1 2227 Intron XP_016855614.1
XM_017000126.1 2227 Intron XP_016855615.1
XM_017000127.1 2227 Intron XP_016855616.1
XM_017000128.1 2227 Intron XP_016855617.1
XM_017000129.1 2227 Intron XP_016855618.1
XM_017000130.1 2227 Intron XP_016855619.1

View Full Product Details