Product Details

SNP ID
rs76172737
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:182409171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAGAGTAACAGATATCAAGAGT[A/G]GCCCTGAAGATAGAATTCTGGGACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00272 PubMed Links

Gene Details

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

Gene
TEDDM1
Gene Name
transmembrane epididymal protein 1
There are no transcripts associated with this gene.

View Full Product Details