Product Details

SNP ID
rs80265056
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:182412244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTTTGTGTGAAGCCCCTACTTT[C/T]TTATTCCTCAAATAAATGAATTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00272 PubMed Links

Gene Details

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

View Full Product Details