Product Details

SNP ID
rs75034882
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:182601331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGGCATCTTCCCCCATTCAAAT[G/T]ATAAGTTTCATTAGGTAAGGGACTT
Phenotype
MIM: 602514
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RGS16 PubMed Links

Gene Details

Gene
RGS16
Gene Name
regulator of G-protein signaling 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002928.3 Intron NP_002919.3

View Full Product Details