Product Details
- SNP ID
-
rs79196773
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:152212807 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCAAAGCTCTTTAAAAGCTTTTC[A/G]TTATTCCTCAAAGTTTAACCCTCAT
- Phenotype
-
MIM: 616293
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HRNR
PubMed Links
Gene Details
- Gene
- HRNR
- Gene Name
- hornerin
View Full Product Details