Product Details

SNP ID
rs74329088
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:192637642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAACATGAGAAAAAAATGATC[A/G]TTGTTTATTTGAAGCTTGGTGAGTT
Phenotype
MIM: 607190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS13 PubMed Links

Gene Details

Gene
RGS13
Gene Name
regulator of G-protein signaling 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002927.4 233 UTR 5 NP_002918.1
NM_144766.2 233 UTR 5 NP_658912.1

View Full Product Details