Product Details

SNP ID
rs74665067
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:197157747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATATTTCTTTTGTTTTACCTCC[A/G]CTAGTATGGCGAATGTTGCTGTAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB41 PubMed Links

Gene Details

Gene
ZBTB41
Gene Name
zinc finger and BTB domain containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194314.2 4411 UTR 3 NP_919290.2
XM_011509507.2 4411 Intron XP_011507809.1
XM_017001205.1 4411 Intron XP_016856694.1

View Full Product Details