Product Details

SNP ID
rs79777593
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:153437427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACAGAGATTGGTGTCAGGCTCC[A/G]CCAGGAACCACACTGGTAAGGAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A7L2 PubMed Links

Gene Details

Gene
S100A7L2
Gene Name
S100 calcium binding protein A7 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045479.1 Intron NP_001038944.2

View Full Product Details