Product Details

SNP ID
rs79811259
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:202335581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTTTAGAAGAATACACAAAAT[G/T]TTTTATTTCAGCACAATCTTCAATA
Phenotype
MIM: 610538
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2T PubMed Links

Gene Details

Gene
UBE2T
Gene Name
ubiquitin conjugating enzyme E2 T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310326.1 Intron NP_001297255.1
NM_014176.3 Intron NP_054895.1

View Full Product Details